Keep your session open?
Ending In 
Your shopping session has expired.
Your session has expired. For your security, we have logged you out.
Would you like to log in again?

Dioctyldichlorosilane ≥97%

Supplier: BeanTown Chemical

131460-50G 131460-10G
BT131460-50GEA 221.24 USD InStock
BT131460-50G BT131460-10G
Dioctyldichlorosilane ≥97%
Dioctyldichlorosilane
  • Refractive index: 1.456
  • Moisture Sensitive
  • Liquid


Formula: C₁₆H₃₄Cl₂Si
Density: 0.936 g/mL
MDL Number: MFCD01074472
CAS Number: 18416-07-4
UN: 2987
ADR: 8,II
Order Now
Specification Test Results
Assay 97%

 

ORDER

DOCUMENTATION

Q and A

Product Questions and Answers

Enter Certificate Details

Enter the Lot number:

Choose from recent batch/lot numbers:

Or
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your cart, enter a quantity and click Add to Cart.

Original Product:

Description Catalog Number Availability Unit Your Price Price Per Qty