Keep your session open?
Ending In 
Your shopping session has expired.
Your session has expired. For your security, we have logged you out.
Would you like to log in again?

2,3-Dibromopropionyl chloride ≥95%

Supplier: BeanTown Chemical
Synonyms: 2,3-Dibromopropionylchloride

132445-50G 132445-10G 132445-250G
BT132445-10GEA 74.06 USD InStock
BT132445-10G BT132445-50G BT132445-250G
2,3-Dibromopropionyl chloride ≥95%
2,3-Dibromopropionyl chloride
  • RTECS: UG6657500
  • Refractive index: 1.542
  • Moisture Sensitive
  • Liquid


Formula: C₃H₃Br₂ClO
MW: 250.31 g/mol
Boiling Pt: 191…193 °C
Density: 2.181 g/mL
Flash Pt: 66 °C (151 °F)
MDL Number: MFCD00000712
CAS Number: 18791-02-1
EINECS: 242-575-2
ADR: 8,II
Order Now
Specification Test Results
Assay 95%

 

ORDER

DOCUMENTATION

Q and A

Product Questions and Answers

Enter Certificate Details

Enter the Lot number:

Choose from recent batch/lot numbers:

Or
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your cart, enter a quantity and click Add to Cart.

Original Product:

Description Catalog Number Availability Unit Your Price Price Per Qty