Keep your session open?
Ending In 
Your shopping session has expired.
Your session has expired. For your security, we have logged you out.
Would you like to log in again?

Perchloric acid 60.0 - 62.0% ACS

Supplier: BeanTown Chemical

132605-500ML 132605-100ML 132605-2L
BT132605-2LEA 269.5 USD InStock
BT132605-2L BT132605-500ML BT132605-100ML
Perchloric acid 60.0 - 62.0% ACS
Perchloric acid
  • RTECS: SC7500000
  • Liquid


Formula: HClO₄
MW: 100.46 g/mol
Boiling Pt: 203 °C
Melting Pt: –112 °C
Density: 1.535 g/mL
Flash Pt: >110 °C (>230 °F)
MDL Number: MFCD00011325
CAS Number: 7601-90-3
EINECS: 231-512-4
UN: 1873
ADR: 5.1,I
https://us-legacy.vwr.com/stibo/web/std.lang.all/06/22/16640622.jpg
Order Now
Specification Test Results
Assay 60-62%
Color (APHA) ≤10
Residue after ignition ≤0.003%
Silicate & phosphate ≤5ppm
Chloride (Cl) ≤0.001%
Nitrogen compounds (as N) ≤0.001%
Sulfate (SO4) ≤0.001%
Heavy metals (as Pb) ≤1ppm
Iron (Fe) ≤1ppm

 

ORDER

DOCUMENTATION

Q and A

Product Questions and Answers

Enter Certificate Details

Enter the Lot number:

Choose from recent batch/lot numbers:

Or
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your cart, enter a quantity and click Add to Cart.

Original Product:

Description Catalog Number Availability Unit Your Price Price Per Qty