Keep your session open?
Ending In 
Your shopping session has expired.
Your session has expired. For your security, we have logged you out.
Would you like to log in again?

Lanthanum ≥99.9% (trace rare earth metals basis), foil, Thickness 0.1 mm

Supplier: BeanTown Chemical

134035-50X50MM 134035-25X25MM
BT134035-50X50EA 362.47 USD InStock
BT134035-50X50 BT134035-25X25
Lanthanum ≥99.9% (trace rare earth metals basis), foil, Thickness 0.1 mm
Lanthanum
  • Solid


Formula: La
MW: 138.91 g/mol
Boiling Pt: 3464 °C
Melting Pt: 920 °C
Density: 6.19 g/mL
MDL Number: MFCD00011066
CAS Number: 7439-91-0
EINECS: 231-099-0
UN: 3208
ADR: 4.3,II
Order Now
Specification Test Results
Assay 99.9%

 

ORDER

DOCUMENTATION

Q and A

Product Questions and Answers

Enter Certificate Details

Enter the Lot number:

Choose from recent batch/lot numbers:

Or
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your cart, enter a quantity and click Add to Cart.

Original Product:

Description Catalog Number Availability Unit Your Price Price Per Qty