Keep your session open?
Ending In 
Your shopping session has expired.
Your session has expired. For your security, we have logged you out.
Would you like to log in again?

Titanium(III) fluoride

Supplier: BeanTown Chemical

134200-50G 134200-10G
BT134200-10GEA 62.17 USD InStock
BT134200-10G BT134200-50G
Titanium(III) fluoride
Titanium (III) fluoride
  • Air Sensitive, Moisture Sensitive
  • Powder


Formula: F₃Ti
MW: 104.88 g/mol
Density: 3.4 g/mL
MDL Number: MFCD00042544
CAS Number: 13470-08-1
EINECS: 236-732-4
UN: 3260
ADR: 8,II
Order Now

 

ORDER

DOCUMENTATION

Q and A

Product Questions and Answers

Enter Certificate Details

Enter the Lot number:

Choose from recent batch/lot numbers:

Or
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your cart, enter a quantity and click Add to Cart.

Original Product:

Description Catalog Number Availability Unit Your Price Price Per Qty