Keep your session open?
Ending In 
Your shopping session has expired.
Your session has expired. For your security, we have logged you out.
Would you like to log in again?

Dichlorodiphenylgermane ≥98%

Supplier: BeanTown Chemical
Synonyms: Diphenylgermanium dichlorideDichlorodiphenylgermane

134670-5G 134670-1G
BT134670-5GEA 364.56 USD InStock
BT134670-5G BT134670-1G
Dichlorodiphenylgermane ≥98%
Dichlorodiphenylgermane
  • Refractive index: 1.598
  • Moisture Sensitive
  • Liquid


Formula: C₁₂H₁₀Cl₂Ge
Boiling Pt: 100 °C/0.005 mmHg
Density: 1.415 g/mL
Flash Pt: >110 °C (>230 °F)
MDL Number: MFCD00013587
CAS Number: 1613-66-7
EINECS: 216-562-7
UN: 3265
ADR: 8,II
Order Now
Specification Test Results
Assay 98%

 

ORDER

DOCUMENTATION

Q and A

Product Questions and Answers

Enter Certificate Details

Enter the Lot number:

Choose from recent batch/lot numbers:

Or
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your cart, enter a quantity and click Add to Cart.

Original Product:

Description Catalog Number Availability Unit Your Price Price Per Qty