Keep your session open?
Ending In 
Your shopping session has expired.
Your session has expired. For your security, we have logged you out.
Would you like to log in again?

Chloroacetonitrile ≥98%

Supplier: BeanTown Chemical

134730-100G 134730-500G 134730-2.5KG
BT134730-500GEA 218.99 USD InStock
BT134730-500G BT134730-100G BT134730-2.5KG
Chloroacetonitrile ≥98%
Chloroacetonitrile
  • RTECS: AL8225000
  • Refractive index: 1.422
  • Liquid


Formula: C₂H₂ClN
MW: 75.5 g/mol
Boiling Pt: 124…126 °C
Density: 1.194 g/mL
Flash Pt: 54 °C (129 °F)
MDL Number: MFCD00001885
CAS Number: 107-14-2
EINECS: 203-467-0
UN: 2668
ADR: 6.1,I
Order Now
Specification Test Results
Assay 98%

 

ORDER

DOCUMENTATION

Q and A

Product Questions and Answers

Enter Certificate Details

Enter the Lot number:

Choose from recent batch/lot numbers:

Or
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your cart, enter a quantity and click Add to Cart.

Original Product:

Description Catalog Number Availability Unit Your Price Price Per Qty