Keep your session open?
Ending In 
Your shopping session has expired.
Your session has expired. For your security, we have logged you out.
Would you like to log in again?

1,4-Dioxane-[D₈] ≥99% (Isotopic)

Supplier: BeanTown Chemical
Synonyms: 1,4-Dioxane-[²H₈]1,4-Diethylene dioxide-D8Glycolethylether-D81,4-Dioxacyclohexane-D8

136015-1G
BT136015-1GEA 152.53 USD InStock
BT136015-1G
1,4-Dioxane-[D₈] ≥99% (Isotopic)
1,4-Dioxane-[D8]
  • Hygroscopic
  • Liquid


Formula: C₄D₈O₂
MW: 96.04 g/mol
Boiling Pt: 101 °C
Melting Pt: 12 °C
Density: 1.13 g/mL
MDL Number: MFCD00044239
CAS Number: 17647-74-4
EINECS: 241-628-7
UN: 1165
ADR: 3,II
Order Now
Specification Test Results
Assay 99%

 

ORDER

DOCUMENTATION

Q and A

Product Questions and Answers

Enter Certificate Details

Enter the Lot number:

Choose from recent batch/lot numbers:

Or
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your cart, enter a quantity and click Add to Cart.

Original Product:

Description Catalog Number Availability Unit Your Price Price Per Qty