Keep your session open?
Ending In 
Your shopping session has expired.
Your session has expired. For your security, we have logged you out.
Would you like to log in again?

Diisopropyl ether ≥99%

Supplier: BeanTown Chemical
Synonyms: Isopropyl ether

137495-1L
BT137495-1LEA 149.8 USD InStock
BT137495-1L
Diisopropyl ether ≥99%
Diisopropyl ether
  • RTECS: TZ5425000
  • Refractive index: 1.367
  • Liquid


Formula: (CH₃)₂CHOCH(CH₃)₂
MW: 102.18 g/mol
Boiling Pt: 68…69 °C
Melting Pt: –85 °C
Density: 0.725 g/mL
Flash Pt: –29 °C (–20 °F)
MDL Number: MFCD00008880
CAS Number: 108-20-3
EINECS: 203-560-6
UN: 1159
ADR: 3,II
https://us-legacy.vwr.com/stibo/web/std.lang.all/11/45/2301145.jpg
Order Now
Specification Test Results
Assay 99%

 

ORDER

DOCUMENTATION

Q and A

Product Questions and Answers

Enter Certificate Details

Enter the Lot number:

Choose from recent batch/lot numbers:

Or
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your cart, enter a quantity and click Add to Cart.

Original Product:

Description Catalog Number Availability Unit Your Price Price Per Qty