Keep your session open?
Ending In 
Your shopping session has expired.
Your session has expired. For your security, we have logged you out.
Would you like to log in again?

Chlorotriethylsilane ≥98%

Supplier: BeanTown Chemical
Synonyms: Triethylchlorosilane

138990-250G 138990-50G 138990-10G
BT138990-50GEA 141.24 USD InStock
BT138990-50G BT138990-250G BT138990-10G
Chlorotriethylsilane ≥98%
Chlorotriethylsilane
  • Refractive index: 1.43
  • Moisture Sensitive
  • Liquid


Formula: C₆H₁₅ClSi
Boiling Pt: 142…144 °C
Density: 0.898 g/mL
Flash Pt: 30 °C (86 °F)
MDL Number: MFCD00000507
CAS Number: 994-30-9
EINECS: 213-615-6
UN: 2986
ADR: 8,II
Order Now
Specification Test Results
Assay 98%

 

ORDER

DOCUMENTATION

Q and A

Product Questions and Answers

Enter Certificate Details

Enter the Lot number:

Choose from recent batch/lot numbers:

Or
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your cart, enter a quantity and click Add to Cart.

Original Product:

Description Catalog Number Availability Unit Your Price Price Per Qty