Keep your session open?
Ending In 
Your shopping session has expired.
Your session has expired. For your security, we have logged you out.
Would you like to log in again?

Hydrofluoric acid 47 - 51%, Environmental Grade

Supplier: BeanTown Chemical

139450-500ML
BT139450-500MLEA 293.49 USD InStock
BT139450-500ML
Hydrofluoric acid 47 - 51%, Environmental Grade
Hydrofluoric acid
  • RTECS: MW7875000
  • Hygroscopic
  • Liquid, single sub-boiling quartz distillation


Formula: HF
MW: 20.01 g/mol
Density: 1.15 g/mL
MDL Number: MFCD00011346
CAS Number: 7664-39-3
EINECS: 231-634-8
UN: 1790
ADR: 8,II
Order Now
Specification Test Results
Assay 47-51%

 

ORDER

DOCUMENTATION

Q and A

Product Questions and Answers

Enter Certificate Details

Enter the Lot number:

Choose from recent batch/lot numbers:

Or
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your cart, enter a quantity and click Add to Cart.

Original Product:

Description Catalog Number Availability Unit Your Price Price Per Qty