Keep your session open?
Ending In 
Your shopping session has expired.
Your session has expired. For your security, we have logged you out.
Would you like to log in again?

Dicarbonylcyclopentadienylcobalt

Supplier: BeanTown Chemical
Synonyms: Cyclopentadienylcobalt dicarbonylDicarbonylcyclopentadienylcobalt(I)

139560-5G 139560-1G
BT139560-5GEA 179.65 USD InStock
BT139560-5G BT139560-1G
Dicarbonylcyclopentadienylcobalt
Dicarbonylcyclopentadienylcobalt
  • Air Sensitive
  • Liquid


Formula: C₇H₅CoO₂
MW: 180.05 g/mol
Boiling Pt: 139…140 °C/710 mmHg
Flash Pt: 27 °C (81 °F)
MDL Number: MFCD00009589
CAS Number: 12078-25-0
EINECS: 235-139-8
ADR: 6.1,II
Order Now

 

ORDER

DOCUMENTATION

Q and A

Product Questions and Answers

Enter Certificate Details

Enter the Lot number:

Choose from recent batch/lot numbers:

Or
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your cart, enter a quantity and click Add to Cart.

Original Product:

Description Catalog Number Availability Unit Your Price Price Per Qty