Keep your session open?
Ending In 
Your shopping session has expired.
Your session has expired. For your security, we have logged you out.
Would you like to log in again?

Lithium hexafluoroarsenate ≥99%

Supplier: BeanTown Chemical

140645-50G 140645-10G
BT140645-10GEA 105.95 USD InStock
BT140645-10G BT140645-50G
Lithium hexafluoroarsenate ≥99%
Lithium hexafluoroarsenate
  • Hygroscopic
  • Powder


Formula: AsF₆Li
MW: 195.85 g/mol
MDL Number: MFCD00011076
CAS Number: 29935-35-1
EINECS: 249-963-0
UN: 1557
ADR: 6.1,II
Order Now
Specification Test Results
Assay 99%

 

ORDER

DOCUMENTATION

Q and A

Product Questions and Answers

Enter Certificate Details

Enter the Lot number:

Choose from recent batch/lot numbers:

Or
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your cart, enter a quantity and click Add to Cart.

Original Product:

Description Catalog Number Availability Unit Your Price Price Per Qty