Keep your session open?
Ending In 
Your shopping session has expired.
Your session has expired. For your security, we have logged you out.
Would you like to log in again?

Sulfuric acid 0.5 N in water volumetric solution

Supplier: BeanTown Chemical

140830-1L 140830-4L 140830-20L
BT140830-20LEA 167.79 USD InStock
BT140830-20L BT140830-1L BT140830-4L
Sulfuric acid 0.5 N in water volumetric solution
Sulfuric acid
  • Liquid


Formula: H₂SO₄
MW: 98.08 g/mol
MDL Number: MFCD00064589
CAS Number: 7664-93-9
EINECS: 231-639-5
UN: 2796
ADR: 8,II
Order Now

 

ORDER

DOCUMENTATION

Q and A

Product Questions and Answers

Enter Certificate Details

Enter the Lot number:

Choose from recent batch/lot numbers:

Or
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your cart, enter a quantity and click Add to Cart.

Original Product:

Description Catalog Number Availability Unit Your Price Price Per Qty