Keep your session open?
Ending In 
Your shopping session has expired.
Your session has expired. For your security, we have logged you out.
Would you like to log in again?

Tantalum carbide ≥99.5% (trace metals basis)

Supplier: BeanTown Chemical
Synonyms: Tantalum(IV) carbide

140895-250G 140895-10G 140895-50G
BT140895-250GEA 1117.65 USD InStock
BT140895-250G BT140895-10G BT140895-50G
Tantalum carbide ≥99.5% (trace metals basis)
Tantalum carbide
  • -325 Mesh Powder


Formula: CTa
Melting Pt: 3880 °C
MDL Number: MFCD00011255
CAS Number: 12070-06-3
EINECS: 235-118-3
UN: 3178
ADR: 4.1,III
Order Now
Specification Test Results
Assay 99.5%

 

ORDER

DOCUMENTATION

Q and A

Product Questions and Answers

Enter Certificate Details

Enter the Lot number:

Choose from recent batch/lot numbers:

Or
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your cart, enter a quantity and click Add to Cart.

Original Product:

Description Catalog Number Availability Unit Your Price Price Per Qty