Keep your session open?
Ending In 
Your shopping session has expired.
Your session has expired. For your security, we have logged you out.
Would you like to log in again?

Lithium-tert-butanolate ≥99.9% (trace metals basis)

Supplier: BeanTown Chemical
Synonyms: Lithium tert-butoxide

140980-25G
BT140980-25GEA 195.24 USD InStock
BT140980-25G
Lithium-tert-butanolate ≥99.9% (trace metals basis)
Lithium tert-butanolate
  • RTECS: UB8520000
  • Air Sensitive, Moisture Sensitive
  • Powder


Formula: C₄H₉LiO
MDL Number: MFCD00050479
CAS Number: 1907-33-1
EINECS: 217-611-5
UN: 3206
ADR: 4.2,II
Order Now
Specification Test Results
Assay 99.9%

 

ORDER

DOCUMENTATION

Q and A

Product Questions and Answers

Enter Certificate Details

Enter the Lot number:

Choose from recent batch/lot numbers:

Or
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your cart, enter a quantity and click Add to Cart.

Original Product:

Description Catalog Number Availability Unit Your Price Price Per Qty