Keep your session open?
Ending In 
Your shopping session has expired.
Your session has expired. For your security, we have logged you out.
Would you like to log in again?

Silver bromide ≥99.9%

Supplier: BeanTown Chemical
Synonyms: Argentous bromide

141290-5G 141290-25G 141290-100G
BT141290-5GEA 36.16 USD InStock
BT141290-5G BT141290-100G BT141290-25G
Silver bromide ≥99.9%
Silver bromide
  • Light Sensitive
  • Powder


Formula: AgBr
MW: 187.77 g/mol
Melting Pt: 432 °C
Density: 6.473 g/mL
MDL Number: MFCD00003398
CAS Number: 7785-23-1
EINECS: 232-076-8
ADR: 9,III
Order Now
Specification Test Results
Assay 99.9%

 

ORDER

DOCUMENTATION

Q and A

Product Questions and Answers

Enter Certificate Details

Enter the Lot number:

Choose from recent batch/lot numbers:

Or
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your cart, enter a quantity and click Add to Cart.

Original Product:

Description Catalog Number Availability Unit Your Price Price Per Qty