Keep your session open?
Ending In 
Your shopping session has expired.
Your session has expired. For your security, we have logged you out.
Would you like to log in again?

Hydrogen peroxide 29 - 32% (w/w) in water stabilized ACS

Supplier: BeanTown Chemical

141405-4X500ML 141405-50ML 141405-500ML
BT141405-500MLEA 71.39 USD InStock
BT141405-500ML BT141405-50ML BT141405-4X500
Hydrogen peroxide 29 - 32% (w/w) in water stabilized ACS
Hydrogen peroxide
  • RTECS: MX0899000
  • Liquid


Formula: H₂O₂
MW: 34.01 g/mol
MDL Number: MFCD00011333
CAS Number: 7722-84-1
EINECS: 231-765-0
UN: 2014
ADR: 5.1,II
Order Now

 

ORDER

DOCUMENTATION

Q and A

Product Questions and Answers

Enter Certificate Details

Enter the Lot number:

Choose from recent batch/lot numbers:

Or
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your cart, enter a quantity and click Add to Cart.

Original Product:

Description Catalog Number Availability Unit Your Price Price Per Qty