Keep your session open?
Ending In 
Your shopping session has expired.
Your session has expired. For your security, we have logged you out.
Would you like to log in again?

Chloroiodomethane ≥98% stabilized

Supplier: BeanTown Chemical

142000-50G 142000-250G 142000-10G
BT142000-50GEA 130.84 USD InStock
BT142000-50G BT142000-250G BT142000-10G
Chloroiodomethane ≥98% stabilized
Chloroiodomethane
  • Refractive index: 1.582
  • Light Sensitive
  • Liquid


Formula: CH₂ClI
MW: 176.38 g/mol
Boiling Pt: 108…109 °C
Density: 2.422 g/mL
MDL Number: MFCD00001078
CAS Number: 593-71-5
EINECS: 209-804-8
UN: 2810
ADR: 6.1,III
Order Now
Specification Test Results
Assay 98%

 

ORDER

DOCUMENTATION

Q and A

Product Questions and Answers

Enter Certificate Details

Enter the Lot number:

Choose from recent batch/lot numbers:

Or
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your cart, enter a quantity and click Add to Cart.

Original Product:

Description Catalog Number Availability Unit Your Price Price Per Qty