Keep your session open?
Ending In 
Your shopping session has expired.
Your session has expired. For your security, we have logged you out.
Would you like to log in again?

Sodium perrhenate ≥99% (trace metals basis)

Supplier: BeanTown Chemical

142080-1G 142080-25G 142080-5G
BT142080-5GEA 159.92 USD InStock
BT142080-5G BT142080-1G BT142080-25G
Sodium perrhenate ≥99% (trace metals basis)
Sodium perrhenate
  • RTECS: WD3675000
  • Hygroscopic
  • Crystalline


Formula: NaO₄Re
MW: 273.19 g/mol
Melting Pt: 300 °C
Density: 5.39 g/mL
MDL Number: MFCD00014231
CAS Number: 13472-33-8
EINECS: 236-742-9
UN: 1479
ADR: 5.1,II
Order Now
Specification Test Results
Assay 99%

 

ORDER

DOCUMENTATION

Q and A

Product Questions and Answers

Enter Certificate Details

Enter the Lot number:

Choose from recent batch/lot numbers:

Or
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your cart, enter a quantity and click Add to Cart.

Original Product:

Description Catalog Number Availability Unit Your Price Price Per Qty