Keep your session open?
Ending In 
Your shopping session has expired.
Your session has expired. For your security, we have logged you out.
Would you like to log in again?

Phenylacetyl chloride ≥98%

Supplier: BeanTown Chemical

142320-1KG 142320-100G 142320-500G
BT142320-100GEA 38.84 USD InStock
BT142320-100G BT142320-1KG BT142320-500G
Phenylacetyl chloride ≥98%
Phenylacetyl chloride
  • Refractive index: 1.533
  • Moisture Sensitive
  • Liquid


Formula: C₈H₇ClO
MW: 154.6 g/mol
Boiling Pt: 225 °C
Density: 1.169 g/mL
Flash Pt: 102 °C (215 °F)
MDL Number: MFCD00000729
CAS Number: 103-80-0
EINECS: 203-146-5
UN: 2577
ADR: 8,II
Order Now
Specification Test Results
Assay 98%

 

ORDER

DOCUMENTATION

Q and A

Product Questions and Answers

Enter Certificate Details

Enter the Lot number:

Choose from recent batch/lot numbers:

Or
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your cart, enter a quantity and click Add to Cart.

Original Product:

Description Catalog Number Availability Unit Your Price Price Per Qty