Keep your session open?
Ending In 
Your shopping session has expired.
Your session has expired. For your security, we have logged you out.
Would you like to log in again?

Cesium nitrate ≥99.5% (trace metals basis)

Supplier: BeanTown Chemical
Synonyms: Nitric acid cesium salt

142460-250G
BT142460-250GEA 217.71 USD InStock
BT142460-250G
Cesium nitrate ≥99.5% (trace metals basis)
Cesium nitrate
  • RTECS: FL0700000
  • Hygroscopic
  • Crystalline


Formula: CsNO₃
MW: 194.91 g/mol
Melting Pt: 414 °C
Density: 3.69 g/mL
MDL Number: MFCD00064817
CAS Number: 7789-18-6
UN: 1451
ADR: 5.1,III
Order Now
Specification Test Results
Assay 99.5%

 

ORDER

DOCUMENTATION

Q and A

Product Questions and Answers

Enter Certificate Details

Enter the Lot number:

Choose from recent batch/lot numbers:

Or
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your cart, enter a quantity and click Add to Cart.

Original Product:

Description Catalog Number Availability Unit Your Price Price Per Qty