Keep your session open?
Ending In 
Your shopping session has expired.
Your session has expired. For your security, we have logged you out.
Would you like to log in again?

n-Propylmagnesium chloride 1 M in 2-MeTHF (2-Methyltetrahydrofuran)

Supplier: BeanTown Chemical

142525-100ML 142525-500ML
BT142525-100MLEA 71.87 USD InStock
BT142525-100ML BT142525-500ML
n-Propylmagnesium chloride 1 M in 2-MeTHF (2-Methyltetrahydrofuran)
n-Propylmagnesium chloride
  • Air Sensitive, Moisture Sensitive
  • Liquid


Formula: C₃H₇ClMg
Density: 0.896 g/mL
Flash Pt: <–30 °C (<–22 °F)
MDL Number: MFCD00000474
CAS Number: 2234-82-4
ADR: 4.3 (3),II
Order Now

 

ORDER

DOCUMENTATION

Q and A

Product Questions and Answers

Enter Certificate Details

Enter the Lot number:

Choose from recent batch/lot numbers:

Or
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your cart, enter a quantity and click Add to Cart.

Original Product:

Description Catalog Number Availability Unit Your Price Price Per Qty