Keep your session open?
Ending In 
Your shopping session has expired.
Your session has expired. For your security, we have logged you out.
Would you like to log in again?

Phenylarsine oxide ≥97%

Supplier: BeanTown Chemical
Synonyms: OxophenylarsineArzenePAO

142965-5G 142965-25G 142965-1G
BT142965-25GEA 407.17 USD InStock
BT142965-25G BT142965-5G BT142965-1G
Phenylarsine oxide ≥97%
Phenylarsine oxide
  • RTECS: CH8100000
  • Crystalline/Powder


Formula: C₆H₅AsO
MW: 168.02 g/mol
Melting Pt: 145…148 °C
MDL Number: MFCD00001990
CAS Number: 637-03-6
EINECS: 211-275-3
UN: 3465
ADR: 6.1,III
Order Now
Specification Test Results
Assay 97%

 

ORDER

DOCUMENTATION

Q and A

Product Questions and Answers

Enter Certificate Details

Enter the Lot number:

Choose from recent batch/lot numbers:

Or
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your cart, enter a quantity and click Add to Cart.

Original Product:

Description Catalog Number Availability Unit Your Price Price Per Qty