Keep your session open?
Ending In 
Your shopping session has expired.
Your session has expired. For your security, we have logged you out.
Would you like to log in again?

Sulfuric acid ≥92%

Supplier: BeanTown Chemical

143035-100ML 143035-500ML
BT143035-500MLEA 596.49 USD InStock
BT143035-500ML BT143035-100ML
Sulfuric acid ≥92%
Sulfuric acid
  • RTECS: WS5600000
  • Liquid


Formula: H₂SO₄
MW: 98.08 g/mol
Boiling Pt: 290 °C
Melting Pt: 3 °C
Density: 1.84 g/mL
MDL Number: MFCD00064589
CAS Number: 7664-93-9
EINECS: 231-639-5
UN: 1830
ADR: 8,II
Order Now
Specification Test Results
Assay 99.9999%

 

ORDER

DOCUMENTATION

Q and A

Product Questions and Answers

Enter Certificate Details

Enter the Lot number:

Choose from recent batch/lot numbers:

Or
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your cart, enter a quantity and click Add to Cart.

Original Product:

Description Catalog Number Availability Unit Your Price Price Per Qty