Keep your session open?
Ending In 
Your shopping session has expired.
Your session has expired. For your security, we have logged you out.
Would you like to log in again?

Nickel (II) sulfamate hydrate

Supplier: BeanTown Chemical

143040-250G 143040-1KG 143040-50G
BT143040-250GEA 155.8 USD InStock
BT143040-250G BT143040-50G BT143040-1KG
Nickel (II) sulfamate hydrate
Nickel (II) sulphamate hydrate
  • RTECS: QR9275000
  • Crystalline


Formula: H₄N₂NiO₆S₂·xH₂O
MW: 250.86 g/mol (anhy)
MDL Number: MFCD00137261
CAS Number: 13770-89-3
EINECS: 237-396-1
ADR: 9,III
Order Now

 

ORDER

DOCUMENTATION

Q and A

Product Questions and Answers

Enter Certificate Details

Enter the Lot number:

Choose from recent batch/lot numbers:

Or
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your cart, enter a quantity and click Add to Cart.

Original Product:

Description Catalog Number Availability Unit Your Price Price Per Qty