Keep your session open?
Ending In 
Your shopping session has expired.
Your session has expired. For your security, we have logged you out.
Would you like to log in again?

Chloroacetic acid ≥99%

Supplier: BeanTown Chemical

143790-1KG 143790-250G
BT143790-250GEA 24.84 USD InStock
BT143790-250G BT143790-1KG
Chloroacetic acid ≥99%
Chloroacetic acid
  • RTECS: AF8575000
  • Hygroscopic
  • Crystalline


Formula: ClCH₂COOH
MW: 94.5 g/mol
Boiling Pt: 189 °C
Melting Pt: 60…63 °C
Flash Pt: 126 °C (259 °F)
MDL Number: MFCD00002683
CAS Number: 79-11-8
EINECS: 201-178-4
UN: 1751
ADR: 6.1,II
https://us-legacy.vwr.com/stibo/web/std.lang.all/20/30/2302030.jpg
Order Now
Specification Test Results
Assay 99%

 

ORDER

DOCUMENTATION

Q and A

Product Questions and Answers

Enter Certificate Details

Enter the Lot number:

Choose from recent batch/lot numbers:

Or
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your cart, enter a quantity and click Add to Cart.

Original Product:

Description Catalog Number Availability Unit Your Price Price Per Qty