Keep your session open?
Ending In 
Your shopping session has expired.
Your session has expired. For your security, we have logged you out.
Would you like to log in again?

Isobutylene oxide (2,2-dimethyloxirane, 1,2-epoxy-2-methylpropane) ≥99%

Supplier: BeanTown Chemical
Synonyms: Isobutylene oxide

144200-25G 144200-5G 144200-100G
BT144200-25GEA 100.23 USD InStock
BT144200-25G BT144200-100G BT144200-5G
Isobutylene oxide (2,2-dimethyloxirane, 1,2-epoxy-2-methylpropane) ≥99%
Isobutylene oxide (2,2-Dimethyloxirane, 1,2-Epoxy-2-methylpropane)
  • RTECS: TZ3580000
  • Refractive index: 1.375
  • Liquid


Formula: C₄H₈O
Boiling Pt: 50…51 °C
Density: 0.812 g/mL
Flash Pt: –32 °C (–25 °F)
MDL Number: MFCD00066354
CAS Number: 558-30-5
EINECS: 209-193-8
UN: 2924
ADR: 3,II
Order Now
Specification Test Results
Assay 99%

 

ORDER

DOCUMENTATION

Q and A

Product Questions and Answers

Enter Certificate Details

Enter the Lot number:

Choose from recent batch/lot numbers:

Or
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your cart, enter a quantity and click Add to Cart.

Original Product:

Description Catalog Number Availability Unit Your Price Price Per Qty