Keep your session open?
Ending In 
Your shopping session has expired.
Your session has expired. For your security, we have logged you out.
Would you like to log in again?

Aluminium isopropoxide ≥98%

Supplier: BeanTown Chemical
Synonyms: AluminiumtriisopropylateAluminium isopropoxideAluminiumtriisopropoxideAluminiumisopropylate

144270-1KG
BT144270-1KGEA 133.84 USD InStock
BT144270-1KG
Aluminium isopropoxide ≥98%
Aluminium isopropoxide
  • RTECS: BD0975000
  • Moisture Sensitive
  • Granular


Formula: C₉H₂₁AlO₃
MW: 204.25 g/mol
Melting Pt: 128…133 °C
Density: 1.035 g/mL
MDL Number: MFCD00008870
CAS Number: 555-31-7
EINECS: 209-090-8
UN: 3181
ADR: 4.1,II
Order Now
Specification Test Results
Assay 98%

 

ORDER

DOCUMENTATION

Q and A

Product Questions and Answers

Enter Certificate Details

Enter the Lot number:

Choose from recent batch/lot numbers:

Or
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your cart, enter a quantity and click Add to Cart.

Original Product:

Description Catalog Number Availability Unit Your Price Price Per Qty