Keep your session open?
Ending In 
Your shopping session has expired.
Your session has expired. For your security, we have logged you out.
Would you like to log in again?

5-Bromoanthranilic acid ≥98%

Supplier: BeanTown Chemical
Synonyms: 2-Amino-5-bromobenzoic acid

144700-25G 144700-100G 144700-5G
BT144700-100GEA 623.21 USD InStock
BT144700-100G BT144700-5G BT144700-25G
5-Bromoanthranilic acid ≥98%
5-Bromoanthranilic acid
  • RTECS: CB2557670
  • Light Sensitive
  • Powder


Formula: C₇H₆BrNO₂
MW: 216.03 g/mol
Melting Pt: 213…215 °C
MDL Number: MFCD00007823
CAS Number: 5794-88-7
EINECS: 227-338-3
UN: 2811
ADR: 6.1,III
Order Now
Specification Test Results
Assay 98%

 

ORDER

DOCUMENTATION

Q and A

Product Questions and Answers

Enter Certificate Details

Enter the Lot number:

Choose from recent batch/lot numbers:

Or
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your cart, enter a quantity and click Add to Cart.

Original Product:

Description Catalog Number Availability Unit Your Price Price Per Qty