Keep your session open?
Ending In 
Your shopping session has expired.
Your session has expired. For your security, we have logged you out.
Would you like to log in again?

Iodotrimethylsilane ≥97% stabilized

Supplier: BeanTown Chemical
Synonyms: Trimethylsilyl iodideTrimethyliodosilane

144705-5G 144705-25G 144705-100G
BT144705-100GEA 505.6 USD InStock
BT144705-100G BT144705-5G BT144705-25G
Iodotrimethylsilane ≥97% stabilized
Iodotrimethylsilane
  • Refractive index: 1.471
  • Light Sensitive, Moisture Sensitive
  • Liquid


Formula: C₃H₉ISi
MW: 200.09 g/mol
Boiling Pt: 106 °C
Density: 1.406 g/mL
Flash Pt: –2 °C (28 °F)
MDL Number: MFCD00001028
CAS Number: 16029-98-4
EINECS: 240-171-0
UN: 2924
ADR: 3,II
Order Now
Specification Test Results
Assay 97%

 

ORDER

DOCUMENTATION

Q and A

Product Questions and Answers

Enter Certificate Details

Enter the Lot number:

Choose from recent batch/lot numbers:

Or
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your cart, enter a quantity and click Add to Cart.

Original Product:

Description Catalog Number Availability Unit Your Price Price Per Qty