Keep your session open?
Ending In 
Your shopping session has expired.
Your session has expired. For your security, we have logged you out.
Would you like to log in again?

Propiolic acid ≥98%

Supplier: BeanTown Chemical
Synonyms: Acetylenecarboxylic Acid2-Propynoic acid

144815-25G 144815-100G 144815-5G
BT144815-25GEA 109.08 USD InStock
BT144815-25G BT144815-5G BT144815-100G
Propiolic acid ≥98%
Propiolic acid
  • RTECS: UD9300000
  • Refractive index: 1.431
  • Liquid


Formula: C₃H₂O₂
MW: 70.05 g/mol
Boiling Pt: 102 °C/200 mmHg
Melting Pt: 16…18 °C
Density: 1.138 g/mL
Flash Pt: 58 °C (136 °F)
MDL Number: MFCD00004360
CAS Number: 471-25-0
EINECS: 207-437-8
UN: 2920
ADR: 8,II
Order Now
Specification Test Results
Assay 98%

 

ORDER

DOCUMENTATION

Q and A

Product Questions and Answers

Enter Certificate Details

Enter the Lot number:

Choose from recent batch/lot numbers:

Or
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your cart, enter a quantity and click Add to Cart.

Original Product:

Description Catalog Number Availability Unit Your Price Price Per Qty