Keep your session open?
Ending In 
Your shopping session has expired.
Your session has expired. For your security, we have logged you out.
Would you like to log in again?

Zinc(II) telluride ≥99.995% (trace metals basis)

Supplier: BeanTown Chemical

144920-50G 144920-10G
BT144920-10GEA 106.99 USD InStock
BT144920-10G BT144920-50G
Zinc(II) telluride ≥99.995% (trace metals basis)
Zinc (II) telluride
  • Refractive index: 3.56
  • 3-12mm Sintered Lump


Formula: TeZn
MW: 192.99 g/mol
Melting Pt: 1240 °C
Density: 6.34 g/mL
MDL Number: MFCD00043079
CAS Number: 1315-11-3
EINECS: 215-260-2
ADR: 6.1,II
Order Now
Specification Test Results
Assay 99.995%

 

ORDER

DOCUMENTATION

Q and A

Product Questions and Answers

Enter Certificate Details

Enter the Lot number:

Choose from recent batch/lot numbers:

Or
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your cart, enter a quantity and click Add to Cart.

Original Product:

Description Catalog Number Availability Unit Your Price Price Per Qty