Keep your session open?
Ending In 
Your shopping session has expired.
Your session has expired. For your security, we have logged you out.
Would you like to log in again?

Perchloric acid ≥70% ACS

Supplier: BeanTown Chemical

211015-2.5L 211015-500ML
BT211015-2.5LEA 509.63 USD InStock
BT211015-2.5L BT211015-500ML
Perchloric acid ≥70% ACS
Perchloric acid
  • RTECS: SC7500000
  • Liquid


Formula: HClO₄
MW: 100.46 g/mol
Boiling Pt: 203 °C
Melting Pt: –112 °C
Density: 1.664 g/mL
Flash Pt: >110 °C (>230 °F)
MDL Number: MFCD00011325
CAS Number: 7601-90-3
EINECS: 231-512-4
UN: 1873
ADR: 5.1,I
https://us-legacy.vwr.com/stibo/web/std.lang.all/06/22/16640622.jpg
Order Now
Specification Test Results
Assay 69.0-72.0%
Color (APHA) ≤10
Residue after ignition ≤0.003%
Silicate & phosphate ≤0.0005%
Chloride (Cl) ≤0.001%
Sulfate (SO4) ≤0.001%
Nitrogen compounds (as N) ≤0.001%
Heavy metals (as Pb) ≤0.0001%
Iron (Fe) ≤0.0001%

 

ORDER

DOCUMENTATION

Q and A

Product Questions and Answers

Enter Certificate Details

Enter the Lot number:

Choose from recent batch/lot numbers:

Or
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your cart, enter a quantity and click Add to Cart.

Original Product:

Description Catalog Number Availability Unit Your Price Price Per Qty