Keep your session open?
Ending In 
Your shopping session has expired.
Your session has expired. For your security, we have logged you out.
Would you like to log in again?

Aluminum phosphate ≥99.99% (trace metals basis)

Supplier: BeanTown Chemical
Synonyms: Aluminium phosphate

211695-5G
BT211695-5GEA 971.17 USD InStock
BT211695-5G
Aluminum phosphate ≥99.99% (trace metals basis)
Aluminum phosphate
  • RTECS: TB6450000
  • Powder


Formula: AlPO₄
MW: 121.951 g/mol
Density: 2.566 g/mL
MDL Number: MFCD00003430
CAS Number: 7784-30-7
EINECS: 232-056-9
ADR: 8,III
Order Now
Specification Test Results
Assay 99.99%

 

ORDER

DOCUMENTATION

Q and A

Product Questions and Answers

Enter Certificate Details

Enter the Lot number:

Choose from recent batch/lot numbers:

Or
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your cart, enter a quantity and click Add to Cart.

Original Product:

Description Catalog Number Availability Unit Your Price Price Per Qty