Keep your session open?
Ending In 
Your shopping session has expired.
Your session has expired. For your security, we have logged you out.
Would you like to log in again?

Diyttrium trisulfide ≥99.9% (trace rare earth metals basis)

Supplier: BeanTown Chemical
Synonyms: Yttrium (III) sulphideYttrium sulphideYttrium sulfideYttrium (III) sulfide

212460-1G 212460-5G
BT212460-5GEA 279.4 USD InStock
BT212460-5G BT212460-1G
Diyttrium trisulfide ≥99.9% (trace rare earth metals basis)
Diyttrium trisulphide
  • Powder


Formula: S₃Y₂
MW: 274.01 g/mol
MDL Number: MFCD00058806
CAS Number: 12039-19-9
EINECS: 234-887-2
UN: 3134
ADR: 4.3,II
Order Now
Specification Test Results
Assay 99.9%

 

ORDER

DOCUMENTATION

Q and A

Product Questions and Answers

Enter Certificate Details

Enter the Lot number:

Choose from recent batch/lot numbers:

Or
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your cart, enter a quantity and click Add to Cart.

Original Product:

Description Catalog Number Availability Unit Your Price Price Per Qty