Keep your session open?
Ending In 
Your shopping session has expired.
Your session has expired. For your security, we have logged you out.
Would you like to log in again?

Sulfuric acid fuming 20% (free SO₃)

Supplier: BeanTown Chemical
Synonyms: Oleum

213590-100G 213590-500G
BT213590-100GEA 63.53 USD InStock
BT213590-100G BT213590-500G
Sulfuric acid fuming 20% (free SO₃)
Sulfuric acid fuming
  • Hygroscopic
  • Liquid


Formula: H₂O₇S₂ (H₂SO₄ + SO₃)
Density: 1.925 g/mL
MDL Number: MFCD00137792
CAS Number: 8014-95-7
UN: 1831
ADR: 8,I
Order Now

 

ORDER

DOCUMENTATION

Q and A

Product Questions and Answers

Enter Certificate Details

Enter the Lot number:

Choose from recent batch/lot numbers:

Or
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your cart, enter a quantity and click Add to Cart.

Original Product:

Description Catalog Number Availability Unit Your Price Price Per Qty