Keep your session open?
Ending In 
Your shopping session has expired.
Your session has expired. For your security, we have logged you out.
Would you like to log in again?

Hexamethylditin ≥97%

Supplier: BeanTown Chemical

213940-25G 213940-5G 213940-1G
BT213940-25GEA 754.09 USD InStock
BT213940-25G BT213940-5G BT213940-1G
Hexamethylditin ≥97%
Hexamethylditin
  • RTECS: WH8280000
  • Air Sensitive
  • Liquid


Formula: C₆H₁₈Sn₂
MW: 327.63 g/mol
Boiling Pt: 182 °C/756 mmHg
Melting Pt: 23…24 °C
Density: 1.58 g/mL
Flash Pt: 61 °C (142 °F)
MDL Number: MFCD00008277
CAS Number: 661-69-8
EINECS: 211-549-2
UN: 2788
ADR: 6.1,II
Order Now
Specification Test Results
Assay 97%

 

ORDER

DOCUMENTATION

Q and A

Product Questions and Answers

Enter Certificate Details

Enter the Lot number:

Choose from recent batch/lot numbers:

Or
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your cart, enter a quantity and click Add to Cart.

Original Product:

Description Catalog Number Availability Unit Your Price Price Per Qty