Keep your session open?
Ending In 
Your shopping session has expired.
Your session has expired. For your security, we have logged you out.
Would you like to log in again?

Thymolphthalein 0.05% (w/v) in ethanol

Supplier: BeanTown Chemical
Synonyms: 3,3-Bis(4-hydroxy-5-isopropyl-o-tolyl)phthalide3,3-Bis(4-hydroxy-5-isopropyl-2-methylphenyl)isobenzofuran-1(3H)-one

214575-500ML 214575-100ML
BT214575-500MLEA 42.59 USD InStock
BT214575-500ML BT214575-100ML
Thymolphthalein 0.05% (w/v) in ethanol
Thymolphthaleine
  • Liquid


Formula: C₂₈H₃₀O₄
MW: 430.54 g/mol
Melting Pt: 251…253 °C
Density: 0.92 g/mL
MDL Number: MFCD00005909
CAS Number: 125-20-2
UN: 1170
ADR: 3,II
Order Now

 

ORDER

DOCUMENTATION

Q and A

Product Questions and Answers

Enter Certificate Details

Enter the Lot number:

Choose from recent batch/lot numbers:

Or
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your cart, enter a quantity and click Add to Cart.

Original Product:

Description Catalog Number Availability Unit Your Price Price Per Qty