Keep your session open?
Ending In 
Your shopping session has expired.
Your session has expired. For your security, we have logged you out.
Would you like to log in again?

Copper(II) perchlorate hexahydrate

Supplier: BeanTown Chemical

214985-50G 214985-250G
BT214985-250GEA 83.93 USD InStock
BT214985-250G BT214985-50G
Copper(II) perchlorate hexahydrate
Copper (II) perchlorate hexahydrate
  • Hygroscopic
  • Crystalline


Formula: Cl₂CuO₈·6H₂O
MW: 370.54 g/mol
Density: 2.23 g/mL
MDL Number: MFCD00149666
CAS Number: 10294-46-9
EINECS: 237-391-4
UN: 1481
ADR: 5.1,II
Order Now

 

ORDER

DOCUMENTATION

Q and A

Product Questions and Answers

Enter Certificate Details

Enter the Lot number:

Choose from recent batch/lot numbers:

Or
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your cart, enter a quantity and click Add to Cart.

Original Product:

Description Catalog Number Availability Unit Your Price Price Per Qty