Keep your session open?
Ending In 
Your shopping session has expired.
Your session has expired. For your security, we have logged you out.
Would you like to log in again?

B-Tribromoborazine ≥96%

Supplier: BeanTown Chemical

215430-1G 215430-5G
BT215430-5GEA 338.59 USD InStock
BT215430-5G BT215430-1G
B-Tribromoborazine ≥96%
B-Tribromoborazine
  • Moisture Sensitive
  • Powder


Formula: B₃Br₃H₃N₃
Melting Pt: 124…127 °C
MDL Number: MFCD00058924
CAS Number: 13703-88-3
ADR: 8,III
Order Now
Specification Test Results
Assay 96%

 

ORDER

DOCUMENTATION

Q and A

Product Questions and Answers

Enter Certificate Details

Enter the Lot number:

Choose from recent batch/lot numbers:

Or
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your cart, enter a quantity and click Add to Cart.

Original Product:

Description Catalog Number Availability Unit Your Price Price Per Qty