Keep your session open?
Ending In 
Your shopping session has expired.
Your session has expired. For your security, we have logged you out.
Would you like to log in again?

5-Amino-2,2-difluoro-1,3-benzodioxole ≥97%

Supplier: BeanTown Chemical
Synonyms: 2,2-Difluoro-1,3-benzodioxol-5-amine3,4-(Difluoromethylenedioxy)aniline

216015-5G 216015-1G
BT216015-5GEA 178.65 USD InStock
BT216015-5G BT216015-1G
5-Amino-2,2-difluoro-1,3-benzodioxole ≥97%
5-Amino-2,2-difluoro-1,3-benzodioxole
  • Light Sensitive
  • Liquid


Formula: C₇H₅F₂NO₂
MW: 173.12 g/mol
Boiling Pt: 95…97 °C/12 mmHg
MDL Number: MFCD00190144
CAS Number: 1544-85-0
UN: 2810
ADR: 6.1,III
Order Now
Specification Test Results
Assay 97%

 

ORDER

DOCUMENTATION

Q and A

Product Questions and Answers

Enter Certificate Details

Enter the Lot number:

Choose from recent batch/lot numbers:

Or
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your cart, enter a quantity and click Add to Cart.

Original Product:

Description Catalog Number Availability Unit Your Price Price Per Qty