Keep your session open?
Ending In 
Your shopping session has expired.
Your session has expired. For your security, we have logged you out.
Would you like to log in again?

Sodium metaarsenite 0.1 N in water volumetric solution

Supplier: BeanTown Chemical

216065-500ML
BT216065-500MLEA 93.74 USD InStock
BT216065-500ML
Sodium metaarsenite 0.1 N in water volumetric solution
Sodium metaarsenite
  • Liquid


Formula: NaAsO₂
MW: 129.91 g/mol
MDL Number: MFCD00003472
CAS Number: 7784-46-5
EINECS: 232-070-5
UN: 1686
ADR: 6.1,II
Order Now

 

ORDER

DOCUMENTATION

Q and A

Product Questions and Answers

Enter Certificate Details

Enter the Lot number:

Choose from recent batch/lot numbers:

Or
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your cart, enter a quantity and click Add to Cart.

Original Product:

Description Catalog Number Availability Unit Your Price Price Per Qty