Keep your session open?
Ending In 
Your shopping session has expired.
Your session has expired. For your security, we have logged you out.
Would you like to log in again?

Bromoform ≥97% stabilized

Supplier: BeanTown Chemical
Synonyms: Tribromomethane

217730-1KG 217730-5KG 217730-250G
BT217730-250GEA 65.15 USD InStock
BT217730-250G BT217730-5KG BT217730-1KG
Bromoform ≥97% stabilized
Bromoform
  • RTECS: PB5600000
  • Refractive index: 1.595
  • Light Sensitive
  • Liquid


Formula: Br₃CH
MW: 252.73 g/mol
Boiling Pt: 146…150 °C
Melting Pt: 5…8 °C
Density: 2.89 g/mL
MDL Number: MFCD00000128
CAS Number: 75-25-2
EINECS: 200-854-6
UN: 2515
ADR: 6.1,III
https://us-legacy.vwr.com/stibo/web/std.lang.all/78/85/717885.jpg
Order Now
Specification Test Results
Assay 97%

 

ORDER

DOCUMENTATION

Q and A

Product Questions and Answers

Enter Certificate Details

Enter the Lot number:

Choose from recent batch/lot numbers:

Or
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your cart, enter a quantity and click Add to Cart.

Original Product:

Description Catalog Number Availability Unit Your Price Price Per Qty