Keep your session open?
Ending In 
Your shopping session has expired.
Your session has expired. For your security, we have logged you out.
Would you like to log in again?

Dimagnesium silicide ≥99.5% (trace metals basis)

Supplier: BeanTown Chemical
Synonyms: Magnesium silicide

219110-10G 219110-50G
BT219110-50GEA 132.93 USD InStock
BT219110-50G BT219110-10G
Dimagnesium silicide ≥99.5% (trace metals basis)
Dimagnesium silicide
  • Air Sensitive, Moisture Sensitive
  • -20 Mesh Powder


Formula: Mg₂Si
MW: 76.70 g/mol
Melting Pt: 1102 °C
Density: 2 g/mL
EINECS: 245-254-5
UN: 2624
ADR: 4.3,II
Order Now
Specification Test Results
Assay 99.5%

 

ORDER

DOCUMENTATION

Q and A

Product Questions and Answers

Enter Certificate Details

Enter the Lot number:

Choose from recent batch/lot numbers:

Or
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your cart, enter a quantity and click Add to Cart.

Original Product:

Description Catalog Number Availability Unit Your Price Price Per Qty