Keep your session open?
Ending In 
Your shopping session has expired.
Your session has expired. For your security, we have logged you out.
Would you like to log in again?

Erbium ≥99.9% (trace rare earth metals basis), pieces

Supplier: BeanTown Chemical

220295-10G
BT220295-10GEA 115.28 USD InStock
BT220295-10G
Erbium ≥99.9% (trace rare earth metals basis), pieces
Erbium
  • Solid


Formula: Er
MW: 167.26 g/mol
Boiling Pt: 2868 °C
Melting Pt: 1529 °C
Density: 9.062 g/mL
MDL Number: MFCD00010987
CAS Number: 7440-52-0
EINECS: 231-160-1
ADR: 4.1,II
Order Now
Specification Test Results
Assay 99.9%

 

ORDER

DOCUMENTATION

Q and A

Product Questions and Answers

Enter Certificate Details

Enter the Lot number:

Choose from recent batch/lot numbers:

Or
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your cart, enter a quantity and click Add to Cart.

Original Product:

Description Catalog Number Availability Unit Your Price Price Per Qty