Keep your session open?
Ending In 
Your shopping session has expired.
Your session has expired. For your security, we have logged you out.
Would you like to log in again?

Diyttrium trioxide ≥99.9% (trace rare earth metals basis) Aerosol Refractory Paint

Supplier: BeanTown Chemical
Synonyms: Yttrium (III) oxideYttrium oxide

220970-13OZ
BT220970-13OZEA 397.81 USD InStock
BT220970-13OZ
Diyttrium trioxide ≥99.9% (trace rare earth metals basis) Aerosol Refractory Paint
Diyttrium trioxide
  • Air Sensitive
  • Aerosol Spray Can


Formula: Y₂O₃
MW: 225.81 g/mol
MDL Number: MFCD00011473
CAS Number: 1314-36-9
ADR: 2.1,
Order Now
Specification Test Results
Assay 99.9%

 

ORDER

DOCUMENTATION

Q and A

Product Questions and Answers

Enter Certificate Details

Enter the Lot number:

Choose from recent batch/lot numbers:

Or
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your cart, enter a quantity and click Add to Cart.

Original Product:

Description Catalog Number Availability Unit Your Price Price Per Qty