Keep your session open?
Ending In 
Your shopping session has expired.
Your session has expired. For your security, we have logged you out.
Would you like to log in again?

Silver (I) sulfide ≥99.9% (trace metals basis)

Supplier: BeanTown Chemical
Synonyms: Disilver sulphideDisilver sulfide

221465-5G
BT221465-5GEA 67.95 USD InStock
BT221465-5G
Silver (I) sulfide ≥99.9% (trace metals basis)
Silver (I) sulphide
  • Powder


Formula: Ag₂S
MW: 247.8 g/mol
Melting Pt: 845 °C
Density: 7.234 g/mL
MDL Number: MFCD00003406
CAS Number: 21548-73-2
EINECS: 244-438-2
ADR: 9,III
Order Now
Specification Test Results
Assay 99.9%

 

ORDER

DOCUMENTATION

Q and A

Product Questions and Answers

Enter Certificate Details

Enter the Lot number:

Choose from recent batch/lot numbers:

Or
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your cart, enter a quantity and click Add to Cart.

Original Product:

Description Catalog Number Availability Unit Your Price Price Per Qty