Keep your session open?
Ending In 
Your shopping session has expired.
Your session has expired. For your security, we have logged you out.
Would you like to log in again?

(±)-2-Methyltetrahydrofuran ≥99% stabilized

Supplier: BeanTown Chemical

223785-500ML 223785-2.5L 223785-100ML
BT223785-2.5LEA 418.21 USD InStock
BT223785-2.5L BT223785-100ML BT223785-500ML
(±)-2-Methyltetrahydrofuran ≥99% stabilized
(±)-2-Methyltetrahydrofuran
  • RTECS: LU2800000
  • Refractive index: 1.406
  • Liquid


Formula: C₅H₁₀O
MW: 86.13 g/mol
Boiling Pt: 78…80 °C
Density: 0.86 g/mL
Flash Pt: –12 °C (10 °F)
MDL Number: MFCD00005367
CAS Number: 96-47-9
EINECS: 202-507-4
UN: 2536
ADR: 3,II
Order Now
Specification Test Results
Assay 99%

 

ORDER

DOCUMENTATION

Q and A

Product Questions and Answers

Enter Certificate Details

Enter the Lot number:

Choose from recent batch/lot numbers:

Or
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your cart, enter a quantity and click Add to Cart.

Original Product:

Description Catalog Number Availability Unit Your Price Price Per Qty