Keep your session open?
Ending In 
Your shopping session has expired.
Your session has expired. For your security, we have logged you out.
Would you like to log in again?

Cerium ≥99% (trace rare earth metals basis), ingot

Supplier: BeanTown Chemical

224675-250G 224675-1KG
BT224675-250GEA 126.73 USD InStock
BT224675-250G BT224675-1KG
Cerium ≥99% (trace rare earth metals basis), ingot
Cerium
  • Solid, packaged under mineral oil


Formula: Ce
MW: 140.12 g/mol
Boiling Pt: 3443 °C
Melting Pt: 795 °C
Density: 6.67 g/mL
MDL Number: MFCD00010924
CAS Number: 7440-45-1
EINECS: 231-154-9
UN: 1333
ADR: 4.1,II
Order Now
Specification Test Results
Assay 99%

 

ORDER

DOCUMENTATION

Q and A

Product Questions and Answers

Enter Certificate Details

Enter the Lot number:

Choose from recent batch/lot numbers:

Or
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your cart, enter a quantity and click Add to Cart.

Original Product:

Description Catalog Number Availability Unit Your Price Price Per Qty